Anti-PRAME Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-7004R-A350
BOSSBS-7004R-A350EA 893 EUR
BOSSBS-7004R-A350
Anti-PRAME Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Functions as a transcriptional repressor, inhibiting the signaling of retinoic acid through the retinoic acid receptors RARA, RARB and RARG. Prevents retinoic acid-induced cell proliferation arrest, differentiation and apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRAME
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR