Anti-Retinoic acid induced 17 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
BS-7709R-A750
BOSSBS-7709R-A750EA 893 EUR
BOSSBS-7709R-A750
Anti-Retinoic acid induced 17 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Increases ligand-dependent transcriptional activity of AR and promotes AR sumoylation. The stimulation of AR activity is dependent upon sumoylation.Tissue specificity:Expressed most abundantly in ovary and, at lower levels, in prostate, spleen and testis. Weak expression, if any, in thymus, small intestine, colon and peripheral blood leukocytes.

Type: Primary
Antigen: Retinoic acid induced 17
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/1067
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR