Anti-GPSM1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-7865R-CY5
BOSSBS-7865R-CY5EA 900 EUR
BOSSBS-7865R-CY5
Anti-GPSM1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Guanine nucleotide dissociation inhibitor (GDI) which functions as a receptor-independent activator of heterotrimeric G-protein signaling. Keeps G(i/o) alpha subunit in its GDP-bound form thus uncoupling heterotrimeric G-proteins signaling from G protein-coupled receptors. Controls spindle orientation and asymmetric cell fate of cerebral cortical progenitors. May also be involved in macroautophagy in intestinal cells. May play a role in drug addiction.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPSM1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR