Anti-TLR5 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-8539R
BOSSBS-8539REA 725 EUR
BOSSBS-8539R
Anti-TLR5 Rabbit Polyclonal Antibody
Antibodies
Luciferase from the firefly has become one of the more widely used reporter proteins for the study of gene expression. Luciferase catalyzes a bioluminescent reaction which requires the substrate luciferin as well as Mg2+ and ATP. Mixing these reagents with the cell extract containing luciferase, results in a flash of light that decays rapidly. This light can be detected by a luminometer. The total light emission is proportional to the luciferase activity of the sample.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: TLR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
More Product Information
Order Now

SPECIFICATIONS

Antigen symbol TLR5
Environmentally Preferable
Antigen name Toll-like Receptor 5
Antigen synonyms TIL3|Toll-like receptor 5|Toll/interleukin-1 receptor-like protein 3
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Multiple
Host Rabbit
Gene ID 7100
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100 μg/ml BSA, 50% glycerol and 0,09% sodium azide. Store at –20 °C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR