Anti-KRT1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-9080R-CY5.5
BOSSBS-9080R-CY5.5EA 900 EUR
BOSSBS-9080R-CY5.5
Anti-KRT1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. It can phosphorylate a large number of proteins. Participates in Wnt signaling.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KRT1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR