Anti-TNKS Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Supplier: Bioss
BS-9104R-A647
BOSSBS-9104R-A647EA 893 EUR
BOSSBS-9104R-A647
Anti-TNKS Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Poly-ADP-ribosyltransferase involved in various processes such as Wnt signaling pathway, telomere length and vesicle trafficking. Acts as an activator of the Wnt signaling pathway by mediating poly-ADP-ribosylation (PARsylation) of AXIN1 and AXIN2, 2 key components of the beta-catenin destruction complex: poly-ADP-ribosylated target proteins are recognized by RNF146, which mediates their ubiquitination and subsequent degradation. Also mediates PARsylation of BLZF1 and CASC3, followed by recruitment of RNF146 and subsequent ubiquitination. Mediates PARsylation of TERF1, thereby contributingMore Product Information