Anti-SASS6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Supplier: Bioss
BS-9301R-A350
BOSSBS-9301R-A350EA 893 EUR
BOSSBS-9301R-A350
Anti-SASS6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
SAS-6 (spindle assembly abnormal protein 6 homolog, HsSAS-6) is a 657 amino acid protein encoded by the human gene SAS6. SAS-6 is a component of the centrosome that contains one PISA (present in SAS-6) domain. LK4, SAS-6, CPAP and other centriole related proteins are required at different stages of procentriole formation and were associated with different centriolar structures. SAS-6 associates only transiently with nascent procentrioles, whereas CEP135 and CPAP form a core structure within the proximal lumen of both parental and nascent centrioles. SAS-6 is necessary for procentrioleMore Product Information