Anti-NEK9 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-9318R-CY3
BOSSBS-9318R-CY3EA 1350 EUR
BOSSBS-9318R-CY3
Anti-NEK9 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Pleiotropic regulator of mitotic progression, participating in the control of spindle dynamics and chromosome separation. Phosphorylates different histones, myelin basic protein, beta-casein, and BICD2. Phosphorylates histone H3 on serine and threonine residues and beta-casein on serine residues. Important for G1/S transition and S phase progression.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NEK9
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR