Self-Laminating Cryogenic Polyester Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Supplier: Brady
170882 170877 170767 170765 170875 170766 170768 170769 170878 170879 170880 170881 170883
BRDY170882EA 71 EUR
BRDY170882 BRDY170877 BRDY170767 BRDY170765 BRDY170875 BRDY170766 BRDY170768 BRDY170769 BRDY170878 BRDY170879 BRDY170880 BRDY170881 BRDY170883
Self-Laminating Cryogenic Polyester Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Labels
Securely label laboratory tubes and vials for cryogenic storage with this self-laminating, ultra-thin polyester label material (B-461). Print almost anywhere with a compact Brady portable label printer.
  • For use on smooth, curved surfaces
  • Withstands extreme temperatures from −196 to +130 °C
  • Chemical-resistant
  • Transparent overlaminate protects print
Versatile labels are ideal for curved surfaces such as vials, centrifuge tubes, test tubes, straws, and slides. Self-Laminating Cryogenic Polyester Labels are able to withstand extreme temperatures from −196More Product Information