Brady® StainerBondz™ Chemical-Resistant Polyester Slide Labels with 76 mm Core
Supplier: Brady
321703
BRDY321703EA 316.1 EUR
BRDY321703
Brady® StainerBondz™ Chemical-Resistant Polyester Slide Labels with 76 mm Core
Labels
Super chemical resistant polyester (B-488) is designed for use in slide staining processes and other laboratory identification requiring extreme chemical resistance.
  • B-488 performs well in commonly used H&E slide staining processes
  • Excellent resistance to various chemicals, oils and water
  • Operating temperature from ―80 to 130 °C
This B-488 material, combined with R-4300 ribbon, is the recipe for a solution that remains resistant to xylene and other chemicals commonly used in slide staining processes.
No print to wipe off, no buckling of theMore Product Information