Magicard™ 600 Printers
Supplier: Brady
321948 321951 321945 321946 321949 321947 321950 321952
BRDY321948EA 1860 EUR
BRDY321948 BRDY321951 BRDY321945 BRDY321946 BRDY321949 BRDY321947 BRDY321950 BRDY321952
Magicard™ 600 Printers
Printers Document Printers
The Magicard 600 with Wi-Fi delivers faster print speeds, higher quality and optimised security features, and integrates seamlessly with existing infrastructure for secure ID data processing.
  • High print quality - The Magicard 600 offers a wide colour gamut and prints at 600×300 dpi, ideal for photo cards with super sharp text in small fonts
  • Digital Shredding™ - Once data has been used for the print job, it is rendered unreadable, helping to ensure the security of user data
Fraud prevention - The Magicard 600 offers the ability to print a personalisedMore Product Information
Order Now
SPECIFICATIONS
Compatibility CR79, CR80 CR79, CR80
Environmentally Preferable
Connections Ethernet, USB 3.0 Type A, Wi-Fi Ethernet, USB 3.0 Type A, Wi-Fi
Display Monochrome LCD display Monochrome LCD display
Model Magicard™ 600 Uno Magicard™ 600 Duo
Modes Automatic, Manual Automatic, Manual
Power supply 110 - 240 V 110 - 240 V
Print method One-sided printing Double-sided printing
Print speed Multi-colour print speed (per hour): 190
Single colour print speed (per hour): 750
Multi-colour print speed (per hour): 190
Single colour print speed (per hour): 750
Resolution 600×300 dpi 600×300 dpi
Supported OS Mac OS 10.10.x, Mac OS 10.7.x, Windows® 10, Windows® 11 Mac OS 10.10.x, Mac OS 10.7.x, Windows® 10, Windows® 11
Temperature range 10...30 °C 10...30 °C