Bulb pipettes (one mark), BLAUBRAND®, Class AS, USP (individual)
Supplier: Brand
USP30601 USP30602 USP30603 USP30619 USP30616 USP30608 USP30605 USP30606 USP30607 USP30617 USP30618 USP30609 USP30610 USP30611 USP30612 USP30613 USP30614 USP30615
BRNDUSP30601EA 42.5 EUR
BRNDUSP30601 BRNDUSP30602 BRNDUSP30603 BRNDUSP30619 BRNDUSP30616 BRNDUSP30608 BRNDUSP30605 BRNDUSP30606 BRNDUSP30607 BRNDUSP30617 BRNDUSP30618 BRNDUSP30609 BRNDUSP30610 BRNDUSP30611 BRNDUSP30612 BRNDUSP30613 BRNDUSP30614 BRNDUSP30615
Bulb pipettes (one mark), BLAUBRAND®, Class AS, USP (individual)
Pipettes Bulb Pipettes
AR-Glas®.
  • Individual USP certificate
  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in blue enamel
  • Waiting time 5 seconds
DIN EN ISO 648
Certifications: DIN EN ISO 648; DE-M marking.