UniFlex™ Stainless Steel Braided SPX Hoses for Water
Supplier: BROEN-LAB
93V12200700 93V40201000 93V41510700 93V40200700
BROE93V12200700EA 42 EUR
BROE93V12200700 BROE93V40201000 BROE93V41510700 BROE93V40200700
UniFlex™ Stainless Steel Braided SPX Hoses for Water
Tubing Transfer Tubing
BROEN-LAB offers UniFlex™ hoses and flexible stainless steel pipes as an easy way to make the installation in the laboratory where safety and efficiency are required.
  • All O-rings are lubricated with silicone based lubricant
  • SPX hose with braiding in stainless steel
  • Braiding: Stainless steel with one green stripe
  • Materials with media contact: SPX, brass, EPDM with silicone based lubricant
  • Sealing: EPDM, Aramid fiber/NBR
  • Minimum length with connections: 160 mm
BROEN-LAB UniFlex™ hoses are the ideal solution where there is a higher requirement forMore Product Information