HCCA matrix
Supplier: BRUKER DALTRONIK
8255344
BRUK8255344EA 954 EUR
BRUK8255344
HCCA matrix
Protein Purification Protein Purification Reagents
Pre-portioned HCCA matrix provides easy and convenient preparation of HCCA matrix solutions.
  • Stable and easy to handle for sample preparation
HCCA has been developed especially for Bruker MALDI instruments and purified to ensure accurate results and maintain system cleanliness. This reduces frequency of preventive maintenance cleans required.