R-12/15, Swing-out rotor for conical bottom centrifuge tube - Dimensions (Ø × length): 17×120mm - Capacity12 - Tube volume 15 ml - Max. speed 4,200 rpm - Max. RCF: LMC-3000 --> 1,700 × g - Tube Manufacturers: Falcon, Greiner bio-one, Sarstead, Cornin 1 * 1 items
Supplier: BIOSAN
R-12/15, Swing-out rotor for conical bottom centrifuge tube - Dimensions (Ø × length): 17×120mm - Capacity12 - Tube volume 15 ml - Max. speed 4,200 rpm - Max. RCF: LMC-3000 --> 1,700 × g - Tube Manufacturers: Falcon, Greiner bio-one, Sarstead, Cornin 1 * 1 items