Automatic solvent extraction systems, rapid, pressurised, SpeedExtractor E-914 and E-916
Supplier: BUCHI
11514042 11514044 11514122 11514124 11516014 11516022 11516024 11516042 11516044
BUCH11514042EA 0 EUR
BUCH11514042 BUCH11514044 BUCH11514122 BUCH11514124 BUCH11516014 BUCH11516022 BUCH11516024 BUCH11516042 BUCH11516044
Automatic solvent extraction systems, rapid, pressurised, SpeedExtractor E-914 and E-916
Extractors Automatic Extractor Systems
SpeedExtractor E-916 / E-914.
  • Extract 6 samples in 30 min and free up your time for other tasks
  • Automated sealing reduces operator intervention and increases process safety
  • Combine extract collection with evaporation, minimize solvent use and avoid costly consumable seals
The SpeedExtractor E-916 / E-914 is an automated instrument for parallel extraction of primarily organic compounds from a variety of solid or semi-solid matrices. Conventional methodologies (e.g. like Soxhlet extraction) are accelerated by using a solvent at elevated temperatures andMore Product Information