Anti-KDM1A Rabbit Polyclonal Antibody

Supplier: Bioworld Technology
BS6189
BWRLBS6189EA 341 EUR
BWRLBS6189
Anti-KDM1A Rabbit Polyclonal Antibody
Antibodies
Recombinant full length Human KDM1A.

M1A pAb detects endogenous levels of M1A protein.

Type: Primary
Antigen: KDM1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR