Anti-HFE Rabbit Polyclonal Antibody

Supplier: Bioworld Technology
BS6621
BWRLBS6621EA 341 EUR
BWRLBS6621
Anti-HFE Rabbit Polyclonal Antibody
Antibodies
Recombinant full length Human HFE.

HFE pAb detects endogenous levels of HFE protein.

Type: Primary
Antigen: HFE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR