Anti-OPTN Rabbit Polyclonal Antibody

Supplier: Cayman Chemical
100002-500
CAYM100002-500EA 418 EUR
CAYM100002-500
Anti-OPTN Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: OPTN
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR