BAM 7 ≥98%

Supplier: Cayman Chemical

Synonyms: 3-Methyl-1-(4-phenyl-2-thiazolyl)-1H-pyrazole-4,5-dione 4-[2-(2-ethoxyphenyl)hydrazone], 4-(2-(2-Ethoxyphenyl)hydrazono)-5-methyl-2-(4-phenylthiazol-2-yl)-2,4-dihydro-3H-pyrazol-3-one, Bax Activator Molecule 7

14566-1 14566-10 14566-25 14566-5
CAYM14566-1EA 43 EUR
CAYM14566-1 CAYM14566-10 CAYM14566-25 CAYM14566-5
BAM 7 ≥98%
BAM 7
Formula: C₂₁H₁₉N₅O₂S
Storage Temperature: Freezer
MDL Number: MFCD01246556
CAS Number: 331244-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR