Anti-5-LO Rabbit Polyclonal Antibody

Supplier: Cayman Chemical
160402-1
CAYM160402-1EA 418 EUR
CAYM160402-1
Anti-5-LO Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: 5-LO
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human, Mouse, Pig, Rat
Order Now

SPECIFICATIONS

Antigen symbol 5-LO
Antigen name 5-Methoxytryptophan
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Hamster,Pig,Human,Rat,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Immunogen human and rat amino acids 130-149; The sequence is 95% homologous to 5-LO from mouse and hamster
Amino acid number 130 to 149
Sequence QHRRKELETRQKQYRWMEWN
Purification purified by affinity chromatography
Cross adsorption No
Storage temperature –20 °C
Shipping temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR