Environmental/HazMat racks for small containers
Supplier: CEMO
8091 8092 7820 7856
CEMG8091EA 572 EUR
CEMG8091 CEMG8092 CEMG7820 CEMG7856
Environmental/HazMat racks for small containers
Furniture Shelves
For storage of water-polluting substances in small containers up to 30 L capacity.
  • With general construction inspection approval no. Z-40.22-388
  • Each level can support up to 200 kg
Environmental/HazMat rack 13/20 is completely hot-dip galvanised and equipped with a 20 L leak-tight tray bottom welded in accordance with StawaR (German guidelines for the requirements of steel container trays of capacity up to 1000 L) lowest storage level.
Environmental/HazMat rack 10/20 is completely galvanised and can be assembled without screws or tools. It has 5More Product Information