Dipotassium oxalate 300 g/l in aqueous solution

Supplier: CUSTOM MADE CHEMICALS LAB
Warning

Synonyms: Oxalic acid potassium salt, Potassiumoxalate, Potassium ethanedioate

FC412791
CMPLFC412791EA 0 EUR
CMPLFC412791
Dipotassium oxalate 300 g/l in aqueous solution
di-Potassium oxalate
Formula: C₂K₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00012464
CAS Number: 583-52-8
EINECS: 209-506-8
Merck Index: 12,07820

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR