Anti-S Tag Rabbit Polyclonal Antibody (Peridinin Chlorophyll)

Supplier: Columbia Biosciences
D11-1831
COBSD11-1831EA 629 EUR
COBSD11-1831
Anti-S Tag Rabbit Polyclonal Antibody (Peridinin Chlorophyll)
Antibodies
Type: Tag
Antigen: S Tag (S tag)
Clonality: polyclonal
Clone:
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR