Anti-DYKDDDDK Tag Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: M2]

Supplier: Columbia Biosciences
D5-1718
COBSD5-1718EA 629 EUR
COBSD5-1718
Anti-DYKDDDDK Tag Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: M2]
Antibodies
Type: Tag
Antigen: DYKDDDDK Tag (DYKDDDDK tag)
Clonality: monoclonal
Clone: M2
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR