Polywipe-C wipes in slide 'n seal bags, PROSAT®
Supplier: CONTEC
PSCS0009B PSCS0012B
CONTPSCS0009BEA 814 EUR
CONTPSCS0009B CONTPSCS0012B
Polywipe-C wipes in slide 'n seal bags, PROSAT®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Slide 'n seal bag is a resealable bag that allows easy dispensing of PROSAT® polywipe-C pre-saturated wipes. The plastic piece glides across the top of the bag ensuring a tight seal every time the bag is closed. No adhesives are used to keep the bag closed.
  • Bulk packed in gusseted slide 'n seal bags; tight seal maintains saturation levels for longer periods of use
  • No adhesive used to close the bag; plastic piece glides across bag top ensuring a tight seal
  • Bag designed to be opened and closed easily, eliminating the waste of unused wipes
  • Knitted wipes