Charging stand for Corning® Stripettor Ultra
Supplier: Corning
4998
CORN4998EA 59.95 EUR
CORN4998
Charging stand for Corning® Stripettor Ultra
Pipette Stands
Charging stand with three positions: Break, rest and charge.