Blanking nuts
Supplier: COWIE
PC702.075 PC702.025 PC702.050 PC702.100 PC702.150
COWIPC702.075EA 12.7 EUR
COWIPC702.075 COWIPC702.025 COWIPC702.050 COWIPC702.100 COWIPC702.150
Blanking nuts
Fixings Nuts
PTFE, white.
  • Chemically inert
  • Reduce contamination and simplify cleaning
  • Non-scratch
NPT blanking nuts/plugs are used to seal unused ports. To assemble simply screw the NPT blanking nut into a corresponding NPT port of the reactor lid.