Remote anchoring system, First-Man-Up™, DBI-SALA®, 3M™
Supplier: CAPITAL SAFETY
2100094
CPSD2100094EA 0 EUR
CPSD2100094
Remote anchoring system, First-Man-Up™, DBI-SALA®, 3M™
Fall Protection Fall Lanyards and Ropes
The telescopic rod connector allows to attach a rope life line above the user, enabling the user to ascend a pylon or to reach the top of a pole in complete safety.
  • Rope life line up to 8 m
Certifications: EN 795, Class B