Centrifuge tubes, with screw caps, EUROTUBO®
Supplier: DELTALAB
401402 301403 301402 401403
DELT401402EA 726 EUR
DELT401402 DELT301403 DELT301402 DELT401403
Centrifuge tubes, with screw caps, EUROTUBO®
Tubes Centrifuge Tubes
Made of PP, translucent or PS, clear, with green HDPE screw cap.
  • PP: autoclavable
  • PP: Withstand forces up to 15000×g
  • PS: Withstand forces up to 7500×g
The external skirt allows the tubes to remain free-standing. Supplied screwed. The sterile model (ethylene oxide) is supplied individually in flow-pack bag, with indication of batch, expiration date, etc. according to the guidelines for sterile products.
Caution: For autoclaving, the cap should be loose on the thread and not tightly fitted.