He-Ne laser
Supplier: DIDALAB
POD013213 POD013210
DIDAPOD013213EA 1190 EUR
DIDAPOD013213 DIDAPOD013210
He-Ne laser
Educational Materials Physics Educational Materials Optics Learning Activities
High quality professional gas laser, Class II
  • Max Power:<1 mW
  • Built-in tube power supply
  • Divergence <2 mrad for unpolarised laser
Delivery information: Supplied with rod Ø 10×130 mm for fitting to optical bench and mains power adapter