Station for Tough-Tags
Supplier: DIVERSIFIED BIOTECH
TTSA-9000
DIVETTSA-9000EA 196 EUR
DIVETTSA-9000
Station for Tough-Tags
Label Holders
Tough tag station holds up to six rolls of Tough-tags®, Tough-spots® and Cryo-tags® labels. The ledge in the station is used to write on labels and the straight edge is to tear off the labels.
  • Built-in cutting edge
  • Convenient writing surface
  • Holds multiple rolls for easy transport