TASKI® Sprint 200 Pur-Eco E1b Multi Purpose Cleaner
Supplier: Diversey
7515239
DIVY7515239EA 31 EUR
DIVY7515239
TASKI® Sprint 200 Pur-Eco E1b Multi Purpose Cleaner
Cleaning Agents Detergents and Surface Cleansers
Alcohol-based daily multi purpose cleaner for streak-free cleaning of all water resistant hard surfaces.
  • pH neutral at in use dilution
  • Fast acting, fast drying
  • Based on an effective combination of alcohol and surfactants derived from natural vegetable sources to minimise the impact on the environment and ensure safer use
  • Unified fresh fragrance throughout the Pur-Eco range
Benefits: Suitable for use on water-resistant hard surfaces, including windows, mirrors, glazed ceramics as well as plastic surfaces. Leaves surfaces free of marks and streaks.More Product Information