BrightVision+ Anti-IgG (Poly-AP)
Supplier: IMMUNOLOGIC
DPVB110AP
DPVB110APEA 1070 EUR
DPVB110AP
BrightVision+ Anti-IgG (Poly-AP)
Antibodies
BrightVision+ histostaining reagents utilise a novel controlled polymerisation technology to prepare polymeric APlinker antibody conjugates. Comparing to conventional biotin-streptavidin based detection kits, BrightVision+ histostaining kits have the advantages of higher amplification power, biotin free and more consistent immunostaining outcomes on archival tissues and on difficult-to-work antibodies.
  • High quality secondary antibody for IHC applications, AP labelled
  • Antigen specificity: Anti-Mouse IgG (H+L), Anti-Rabbit IgG (H+L)
  • Kit contains post-