Classic door technology (door handle) For the storage and charging of lithium-ion batteries/rechargeable batteries Type examination by MPA/TÜV SÜD in accordance with DIN EN 14470-1, DIN EN 16121, DIN EN 16122, DIN 31000 and IEC 61439 Risk prevention 1 * 1 items
Supplier: DUPERTHAL
Classic door technology (door handle) For the storage and charging of lithium-ion batteries/rechargeable batteries Type examination by MPA/TÜV SÜD in accordance with DIN EN 14470-1, DIN EN 16121, DIN EN 16122, DIN 31000 and IEC 61439 Risk prevention 1 * 1 items