Tyvek IsoClean IC 270B WS Labcoat (Sterile-only, Double bagged). Bound seams. Covered elastication at wrists. Front snap closure for easy donning and doffing. Packed in a dual barrier validated packaging  system (double bagged). White. Quantity per s 1 * 30 items
Supplier: DuPont