Hooded coverall. Tyvek at the front and large breathable SMS at the back. Stitched seams which are also taped on the upper body: underarms and wrists where Tyvek under gloves are attached. Attached Tyvek socks. Elasticated hood. Elasticated waist (st 1 * 25 items
Supplier: DuPont