Chemical resistant gowns, Tychem® 2000 C and Tychem® 6000 F, model PL50
Supplier: DuPont
TYCPL50SYLLXXL TF0290TGY00L2X TF0290TGY00SM TF0290TGY0034X
113-0876EA 0 EUR
113-0876 DUPOTF0290TGY00L2X DUPOTF0290TGY00SM DUPOTF0290TGY0034X
Chemical resistant gowns, Tychem® 2000 C and Tychem® 6000 F, model PL50
Disposable Garments
Shin-length gown with wrap-over rear closure, hook and loop neck closure, and waist ties.
  • Elasticated wrists
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Anti-static treatment (EN 1149-5) on inside
Tychem® 2000 C garments utilise the strength of Tyvek® and a polymeric barrier coating to offer good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure). They are used for splash or pressurised splash protection in a variety of industrial environments,More Product Information