Chemical resistant overalls, Tyvek® 200 Easysafe
Supplier: DuPont
TSCHF5SWHLG010 TSCHF5SWHMD010 TSCHF5SWHXL010 TSCHF5SWH2X010 TSCHF5SWHSM010 TSCHF5SWH3X010
DUPOTSCHF5SWHLG010EA 732 EUR
DUPOTSCHF5SWHLG010 DUPOTSCHF5SWHMD010 DUPOTSCHF5SWHXL010 DUPOTSCHF5SWH2X010 DUPOTSCHF5SWHSM010 DUPOTSCHF5SWH3X010
Chemical resistant overalls, Tyvek® 200 Easysafe
Disposable Garments
These hooded overalls are made of Easysafe fabric, an optimised non woven PE fabric from DuPont. It is softer and with a more open structure than Tyvek®. It is therefore less protective than Tyvek® (L1431N), but offers adequate Type 5 and 6 barrier protection and remains both air and moisture vapour permeable.
  • Robust yet lightweight (<180 g per garment)
  • Two-piece hood, zip closure with flap
  • Elasticated wrists, ankles and face
  • Elasticated waist (stitched-in)
  • Antistatic treatment (EN 1149-1) - on outside
  • Stitched external seams in yellow for visual
More Product Information
Order Now
SPECIFICATIONS
Model 200 Easysafe
Environmentally Preferable
Anti-static +
Gender
Garment application Chemical protection,Laboratory,Industrial
Weight <180 g