Guanidinium hydrochloride ≥98.5%, Technical Grade low ash residue after ignition
Supplier: VWR Chemicals
Warning
Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
E424-500GEA 201 EUR
E424-500G
Guanidinium hydrochloride ≥98.5%, Technical Grade low ash residue after ignition
Guanidinium hydrochloride
Denaturant that does not contain anti-caking agent.
Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Ammonium Chloride <0.5%
Ash <0.4%
Moisture (KF) <0.3%
Purity >98.5%