Loggers for RDG Validation, SL2001 Set: EBI 12-T220, EBI 12-T441, EBI 12-TP231
Supplier: EBRO
1340-6068 1250-2001
EBRO1340-6068EA 6690 EUR
EBRO1340-6068 EBRO1250-2001
Loggers for RDG Validation, SL2001 Set: EBI 12-T220, EBI 12-T441, EBI 12-TP231
Data Loggers
The SL2001 set by ebro, including the EBI 12-T220, EBI 12-T441, and EBI 12-TP231 loggers, is tailored for validation of washer-disinfectors (RDG) in healthcare and industrial settings, ensuring compliance with cleaning and disinfection standards.
  • Complete validation set for washer-disinfectors
  • Includes EBI 12-T220, EBI 12-T441, and EBI 12-TP231 loggers
  • Monitors temperature and pressure with high precision
  • Ensures hygiene compliance in healthcare and industrial sectors
  • Robust, IP-rated devices for durability in harsh environments
  • Factory calibration