Hightextra® Smooth Wipes
Supplier: ECOLINE NV SA
HTS0060R05300W
ECOLHTS0060R05300WEA 104 EUR
ECOLHTS0060R05300W
Hightextra® Smooth Wipes
Wipes Reusable Cleaning Cloths Cloths for General Applications
The combination of woodpulp and continuous polyester fibres creates a super strong wipe, even when used wet, that copes with tougher cleaning too.