Experimentation kits, principles of thin layer chromatography
Supplier: EDVOTEK
113
EDVO113EA 138 EUR
EDVO113
Experimentation kits, principles of thin layer chromatography
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
This experiment introduces chromatographic theory and methods of thin layer chromatography (TLC). A mixture of dyes are separated on a cellulose-based TLC plate using different solvent systems.
  • For 8 lab groups
  • Spotting plates 20 minutes, TLC separation 5 minutes
Also required: 5ml or 10 ml pipettes.
Delivery information: Kit includes instructions, samples, reagents and solvents, cellulose TLC plate, 5 µl capillary pipettes.