TAE buffer, 50X
Supplier: EDVOTEK
607
EDVO607EA 33 EUR
EDVO607
TAE buffer, 50X
Buffers Electrophoresis Buffers Running Buffers
This 50X concentrated solution (Tris-acetate, EDTA, pH 7,8) is sufficient for making 5 L of diluted working buffer for making agarose gels and electrophoresis chamber buffer.