SYBR® Safe Stain
Supplier: EDVOTEK
608
EDVO608EA 100 EUR
EDVO608
SYBR® Safe Stain
Stains Electrophoresis Stains
SYBR® Safe Stain is a DNA stain that fluoresces with a bright green colour when excited with UV light. Like ethidium bromide, SYBR® Safe Stain binds specifically to the DNA double helix.
  • SAFE for the Biotechnology Classroom
  • More sensitive than ethidium bromide
  • Non-mutagenic
  • 10,000 X Concentrate for 750 ml
  • Volume: 80 µl
Caution: Store in dark at room temperature over 25 °C.