PCR beads
Supplier: EDVOTEK
625
EDVO625EA 53.9 EUR
EDVO625
PCR beads
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Each PCR Bead contains Taq DNA polymerase, Taq DNA polymerase buffer, dNTP mixture and MgCl₂. Pack contains sufficient for 25 reactions.