Benzonitrile

Supplier: EHRENSTORFER
Warning

Synonyms: Cyanobenzene, Phenylcyanide

C10538500
EHERC10538500EA 39.7 EUR
EHERC10538500
Benzonitrile
Benzonitrile
Formula: C₇H₅N
MW: 103.12 g/mol
Boiling Pt: 190.7
Melting Pt: –13 °C
Density: 1 g/cm³ (20 °C)
Flash Pt: 70
Storage Temperature: Ambient
MDL Number: MFCD00001770
CAS Number: 100-47-0
EINECS: 202-855-7
UN: 2224
ADR: 6.1 II
Merck Index: 13,01098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR