Fluorene

Supplier: EHRENSTORFER

Synonyms: 9H-Fluorene, Diphenylenmethane

C20800000
EHERC20800000EA 52.1 EUR
EHERC20800000
Fluorene
Fluorene
Formula: C₁₃H₁₀
MW: 166.22 g/mol
Boiling Pt: 295
Melting Pt: 113…115 °C
Density: 1.203 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001111
CAS Number: 86-73-7
EINECS: 201-695-5
UN: 3077
ADR: 9 III
Merck Index: 13,04181

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR