Baskets and Accessories for Modular Basket Systems
Supplier: ELMA SCHMIDBAUER
1125211 1125224 1125226 1125221 1125212 1125228 3000006012 3000006018 1125217 1124042 1125209 1125214 1125213 1125215 1125220 3000006015 1125210 1125208 1125216 3000009104 1125222 1125223 3000006013 3000006016 3000006017 1125227 3000006014 1125218 1125219 1125225
ELMA1125211EA 15 EUR
ELMA1125211 ELMA1125224 ELMA1125226 ELMA1125221 ELMA1125212 ELMA1125228 ELMA3000006012 ELMA3000006018 ELMA1125217 ELMA1124042 ELMA1125209 ELMA1125214 ELMA1125213 ELMA1125215 ELMA1125220 ELMA3000006015 ELMA1125210 ELMA1125208 ELMA1125216 ELMA3000009104 ELMA1125222 ELMA1125223 ELMA3000006013 ELMA3000006016 ELMA3000006017 ELMA1125227 ELMA3000006014 ELMA1125218 ELMA1125219 ELMA1125225
Baskets and Accessories for Modular Basket Systems
Ultrasonic Baths
These accessories for the modular basket system for Elmasonic P, Elmasonic select, EASY make cleaning small parts easy. These components for use with the Modular Basket System allow users to combine, adapt and customise the basket system for their requirements for cleaning with for Elmasonic P, Elmasonic Select and EASY ultrasonic baths.
  • Basket for cleaning small parts
  • Compatible with Elma pins
  • Modular Basket System is a powerful tool enabling effective and efficient cleaning processes as well as flexible loading
  • The number of objects that can be cleaned