Plate rack, ImmunoSet®
Supplier: ENZO LIFE SCIENCES
ADI801930
ENZOADI801930EA 141 EUR
ENZOADI801930
Plate rack, ImmunoSet®
Microplates Immunology Microplates and Strips
This ImmunoSet® plate rack is a convenient set of five 96 well, clear, high-binding, polystyrene plates and 5 plate sealers.
Use of these plates will provide optimal performance. The use of different types of plates will produce different results and may affect the sensitivity of the assay due to the type of plastic used.